Anti-Neurogenin3 / NGN-3 Mouse Monoclonal Antibody [clone: NGN3/7698]
Catalog # 77180-468
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Neurogenin3 / NGN-3
- Antigen symbol:Neurogenin3 / NGN-3
- Clonality:Monoclonal
- Clone:NGN3/7698
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y4Z2
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:23.54 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human NEUROG3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77180-468
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [NGN3/7698] antibody to Neurogenin3 / NGN-3 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Neurogenin3 / NGN-3
Clonality: Monoclonal
Clone: NGN3/7698
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human