Anti-MGMT Rabbit Monoclonal Antibody [clone: MGMT/8364R]
Catalog # 77180-292
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:6 O methylguanine DNA methyltransferase
- Antigen symbol:MGMT
- Clonality:Monoclonal
- Clone:MGMT/8364R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P16455
- Antigen synonyms:AI267024|Methylated DNA protein cysteine methyltransferase|Agat|MGMT|Methylated-DNA--protein-cysteine methyltransferase|MGC107020|6-O-methylguanine-DNA methyltransferase|EC 2.1.1.63|AGT
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:21.646 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-100) of human MGMT. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77180-292
- Packaging:Plastic vial
Specifications
About this item
Rabbit monoclonal [MGMT/8364R] antibody to MGMT for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: MGMT/8364R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human