Anti-Leptin Receptor Mouse Monoclonal Antibody [clone: LEPR/4545]
Catalog # 77180-174
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:CD 295
- Antigen symbol:Leptin Receptor
- Clonality:Monoclonal
- Clone:LEPR/4545
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P48357
- Antigen synonyms:LEP-R|Db|CD295|HuB219|LEPR|Fa|LEPRD|CD295 antigen|LEP R
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:100 kDa/125 kDa (short/long form)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 335-425) of human LEPR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77180-174
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [LEPR/4545] antibody to Leptin Receptor for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Leptin Receptor
Clonality: Monoclonal
Clone: LEPR/4545
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human