Anti-Estrogen Inducible Protein pS2 Rabbit Recombinant Antibody [clone: TFF1/8817R]
Catalog # 77179-540
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:BCEI
- Antigen symbol:Estrogen Inducible Protein pS2
- Clonality:Recombinant
- Clone:TFF1/8817R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P04155
- Antigen synonyms:Estrogen Inducible Protein pS2|Gastrointestinal trefoil protein pS2|hP1.A|Breast cancer estrogen-inducible protein|D21S21|Breast cancer estrogen inducible sequence|HP1A|Gastrointestinal trefoil protein|Breast cancer estrogen inducible protein
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:6.5 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-84) of human TFF1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77179-540
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [TFF1/8817R] antibody to Estrogen Inducible Protein pS2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Estrogen Inducible Protein pS2
Clonality: Recombinant
Clone: TFF1/8817R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human