Anti-Desmoglein 3 / PVA Rabbit Recombinant Antibody [clone: DSG3/8613R]
Catalog # 77179-466
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Desmoglein 3 / PVA
- Antigen symbol:Desmoglein 3 / PVA
- Clonality:Recombinant
- Clone:DSG3/8613R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P32926
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:130 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 300-500) of human DSG3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77179-466
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [DSG3/8613R] antibody to Desmoglein 3 / PVA for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Desmoglein 3 / PVA
Clonality: Recombinant
Clone: DSG3/8613R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human