Anti-Creatine Kinase MB Rabbit Recombinant Antibody [clone: CKBB/8608R]
Catalog # 77179-326
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:B-CK
- Antigen symbol:Creatine Kinase MB
- Clonality:Recombinant
- Clone:CKBB/8608R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P12277
- Antigen synonyms:creatine kinase muscle|Creatine Kinase MB|creatine kinase B|creatine kinase M|CKMM|brain creatine kinase|CKMB|creatine kinase brain|CKBB
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human full-length CKB protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77179-326
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [CKBB/8608R] antibody to Creatine Kinase MB for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Creatine Kinase MB
Clonality: Recombinant
Clone: CKBB/8608R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human