Anti-Creatine Kinase MB Mouse Recombinant Antibody [clone: rCKBB/8844]
Catalog # 77179-324
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:B-CK
- Antigen symbol:Creatine Kinase MB
- Clonality:Recombinant
- Clone:rCKBB/8844
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P12277
- Antigen synonyms:creatine kinase muscle|Creatine Kinase MB|creatine kinase B|creatine kinase M|CKMM|brain creatine kinase|CKMB|creatine kinase brain|CKBB
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human full-length CKB protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77179-324
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rCKBB/8844] antibody to Creatine Kinase MB for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Creatine Kinase MB
Clonality: Recombinant
Clone: rCKBB/8844
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human