Anti-YOD1 Rabbit Monoclonal Antibody [clone: ARC60044]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC60044] antibody to YOD1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: YOD1
Clonality: Monoclonal
Clone: ARC60044
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77210-126
- Antigen symbol:
- YOD1
- Antigen name:
- DUBA-8
- Antigen synonyms:
- PRO0907|OTUD2|DUBA8|OTU domain containing 2|OTU1_HUMAN|OTU domain-containing protein 2|HsHIN7|HIN-7|HIV-1-induced protease 7
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC60044
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q5VVQ6
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant protein of human YOD1.
- Form:
- Liquid
- Molecular weight:
- 38 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together