Anti-Collagen IV Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Collagen IV for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Collagen IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77213-914
- Antigen symbol:
- Collagen IV
- Antigen name:
- Arresten
- Antigen synonyms:
- RATOR|collagen alpha-1(IV) chain|Collagen IV|CO4A1_HUMAN|BSVD|COL4A1|collagen type IV alpha 1 chain
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# P53420
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant protein of human COL4A4.
- Form:
- Liquid
- Molecular weight:
- 160 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Frequently Bought Together