Anti-TrkA + TrkB + TrkC Rabbit Monoclonal Antibody [clone: ARC59539]
Supplier: Antibodies.com
Certificates
About this item
Rabbit monoclonal [ARC59539] antibody to TrkA + TrkB + TrkC for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: TrkA + TrkB + TrkC
Clonality: Monoclonal
Clone: ARC59539
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77209-980
- Antigen symbol:
- TrkA + TrkB + TrkC
- Antigen name:
- BDNF/NT-3 growth factors receptor
- Antigen synonyms:
- High affinity nerve growth factor receptor|Neurotrophic tyrosine kinase receptor type 3|GP145-TrkB|NT-3 growth factor receptor|MTC|Neurotrophic tyrosine kinase receptor type 1|GP145-TrkC|Neurotrophic tyrosine kinase receptor type 2|gp140trk
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC59539
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P04629, Q16620, Q16288
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthesized peptide derived from human pan-Trk.
- Form:
- Liquid
- Molecular weight:
- 120 - 140 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Specifications
Related Information
Frequently Bought Together