Anti-Interferon beta Rabbit Monoclonal Antibody [clone: ARC58590]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC58590] antibody to Interferon beta for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:2,000 to 1:7,000
Type: Primary
Antigen: Interferon beta
Clonality: Monoclonal
Clone: ARC58590
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77206-408
- Antigen symbol:
- Interferon beta
- Antigen name:
- beta-interferon
- Antigen synonyms:
- IFNB|IFB|Fibroblast interferon|IFNB 1|IFF|IFN-beta|IFNB1|IFN beta|IFNB_HUMAN
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC58590
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P01575
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant protein of human IFN-beta.
- Form:
- Liquid
- Molecular weight:
- 60 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Specifications
Related Information
Frequently Bought Together