Anti-K6-linkage specific Polyubiquitin Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to K6-linkage specific Polyubiquitin for WB and Dot Blot with samples derived from Mouse and Rat.
- Validated Applications: WB, Dot Blot
- Recommended Dilutions: WB: 1:500 to 1:2,000, DB: 1:500 to 1:1,000
Type: Primary
Antigen: K6-linkage specific Polyubiquitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Specifications
- Catalog No:
- 77208-152
- Antigen symbol:
- K6-linkage specific Polyubiquitin
- Antigen name:
- K6-linkage specific Polyubiquitin
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Rat, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence containing polyubiquitinated protein (K6 linkage).
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Tested applications:
- Dot Blot
Specifications
Related Information
Frequently Bought Together