Anti-Granulin Rabbit Monoclonal Antibody [clone: ARC59451]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC59451] antibody to Granulin for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: Granulin
Clonality: Monoclonal
Clone: ARC59451
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77209-938
- Antigen symbol:
- Granulin
- Antigen name:
- Acrogranin
- Antigen synonyms:
- Granulin D|GEP|Granulin E|Granulin|Granulin A|Granulin B|Granulin C|GP88|CLN11
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC59451
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P28799
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant protein of human Granulin.
- Form:
- Liquid
- Molecular weight:
- 70 - 88 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together