Anti-DARC Rabbit Monoclonal Antibody [clone: ARC59631]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC59631] antibody to DARC for ICC/IF with samples derived from Human.
- Validated Applications: ICC/IF
- Recommended Dilutions: ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: DARC
Clonality: Monoclonal
Clone: ARC59631
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77201-420
- Antigen symbol:
- DARC
- Antigen name:
- CCBP1
- Antigen synonyms:
- DUFFY_HUMAN|Fy glycoprotein|Duffy blood group antigen|CD234|Duffy blood group chemokine receptor|DARC|Duffy antigen/chemokine receptor|duffy blood group|Dfy
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC59631
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q16570
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant protein of human DARC.
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF
Specifications
Frequently Bought Together