Anti-Desmoglein 3 / PVA Rabbit Monoclonal Antibody [clone: ARC1190]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1190] antibody to Desmoglein 3 / PVA for WB and ICC/IF with samples derived from Human and Mouse.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Desmoglein 3 / PVA
Clonality: Monoclonal
Clone: ARC1190
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77231-868
- Antigen symbol:
- Desmoglein 3 / PVA
- Antigen name:
- Desmoglein 3 / PVA
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1190
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P32926
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 900-999 of human Desmoglein 3 (P32926).
- Sequence:
- TLSGSQGASALSTSGSVQPAVSIPDPLQHGNYLVTETYSASGSLVQPSTAGFDPLLTQNVIVTERVICPISSVPGNLAGPTQLRGSHTMLCTEDPCSRLI
- Form:
- Liquid
- Molecular weight:
- 145 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF
Specifications
Related Information
Frequently Bought Together