Anti-TRPS1 Rabbit Recombinant Antibody [clone: TRPS1/8131R]
77178-438
- Antibody type:Primary
- Antigen name:GC79
- Antigen symbol:TRPS1
- Clonality:Recombinant
- Clone:TRPS1/8131R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UHF7
- Antigen synonyms:Tricho rhino phalangeal syndrome I|Tricho-rhino-phalangeal syndrome type I protein|Transcriptional repressor GATA binding 1|TRPS1|TRPS 1|TRPS1 gene|Trichorhinophalangeal syndrome I|Trichorhinophalangeal syndrome I homolog|LGCR
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:142 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 900-1100) of human TRPS1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77178-438
- Packaging:Plastic vial
Recombinant rabbit monoclonal [TRPS1/8131R] antibody to TRPS1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TRPS1
Clonality: Recombinant
Clone: TRPS1/8131R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human