Anti-Topoisomerase II alpha Rabbit Recombinant Antibody [clone: TOP2A/7148R]
77178-394
- Antibody type:Primary
- Antigen name:alpha isozyme
- Antigen symbol:Topoisomerase II alpha
- Clonality:Recombinant
- Clone:TOP2A/7148R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P11388
- Antigen synonyms:DNA topoisomerase 2-alpha|DNA topoisomerase II alpha isozyme|DNA gyrase|DNA Topoisomerase2|DNA topoisomerase II 170 kD|DNA topoisomerase (ATP hydrolyzing)|DNA topoisomerase II|DNA topoisomerase 2 alpha|ATP hydrolyzing DNA topoisomerase II alfa
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:170 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Topo II α residues within aa1431-1531 of Topo II α
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77178-394
- Packaging:Plastic vial
Recombinant rabbit monoclonal [TOP2A/7148R] antibody to Topoisomerase II alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Topoisomerase II alpha
Clonality: Recombinant
Clone: TOP2A/7148R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human