Anti-TFE3 Rabbit Recombinant Antibody [clone: TFE3/6849R]
Catalogue # 77178-348
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Antibody type:Primary
- Antigen name:bHLH e33
- Antigen symbol:TFE3
- Clonality:Recombinant
- Clone:TFE3/6849R
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P19532
- Antigen synonyms:bHLHe33|TFE 3|Renal cell carcinoma, papillary|RCCP 2|Tcfe 3|Tcfe3|RCCP2|RCCX1|Class E basic helix-loop-helix protein 33
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:62 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TFE3 protein
- Purification:Protein A/G chromatography
- Cat. no.:77178-348
- Packaging:Plastic vial
Spécifications
À propos de cet article
Recombinant rabbit monoclonal [TFE3/6849R] antibody to TFE3 with samples derived from Human.
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TFE3
Clonality: Recombinant
Clone: TFE3/6849R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human