Anti-Sarcomeric alpha Actinin Mouse Recombinant Antibody [clone: rACTN2/7361]
77178-122
- Antibody type:Primary
- Antigen name:Actin binding protein
- Antigen symbol:Sarcomeric alpha Actinin
- Clonality:Recombinant
- Clone:rACTN2/7361
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35609
- Antigen synonyms:Alpha actinin skeletal muscle isoform 2|Alpha actinin skeletal muscle|ACTN2|Alpha-actinin skeletal muscle isoform 2|Actinin alpha 2|Alpha-actinin-2|Alpha actinin 2|ACTN 2|ACTN2_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:103 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A recombinant fragment (aa557-692) of human ACTN2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77178-122
- Packaging:Plastic vial
Recombinant mouse monoclonal [rACTN2/7361] antibody to Sarcomeric alpha Actinin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Sarcomeric alpha Actinin
Clonality: Recombinant
Clone: rACTN2/7361
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human