Anti-S100 Calcium Binding Protein A13/S100A13 Mouse Monoclonal Antibody [clone: S100A13/7483]
Catalog # 77178-050
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# Q99584
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:10 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-93) of human S100A13 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77178-050
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S100A13/7483] antibody to S100 Calcium Binding Protein A13/S100A13 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: S100 Calcium Binding Protein A13/S100A13
Clonality: Monoclonal
Clone: S100A13/7483
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human