Anti-Osteopontin Mouse Recombinant Antibody [clone: OSP/8693R]
Catalogue # 77177-770
Fournisseur: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Form:Liquid
- Gene ID:UniprotID# P10451
- Antigen synonyms:Bone sialoprotein 1|BSP I|MGC110940|Nephropontin|ETA1|OPN|BSPI|Early T lymphocyte activation 1|ETA 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:34.9 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length rat Spp1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-770
- Packaging:Plastic vial
Spécifications
À propos de cet article
Recombinant mouse monoclonal [OSP/8693R] antibody to Osteopontin for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Canine, Human, Mink, Mouse, Rabbit, Rat and Turkey.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Functional Studies, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Osteopontin
Clonality: Recombinant
Clone: OSP/8693R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Dog, Human, Mink, Mouse, Rabbit, Rat, Turkey