Anti-Mesothelin Mouse Recombinant Antibody [clone: rMSLN/8764]
Catalog # 77177-514
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:CAK 1
- Antigen symbol:Mesothelin
- Clonality:Recombinant
- Clone:rMSLN/8764
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13421
- Antigen synonyms:MPF|CAK1|Megakaryocyte potentiating factor|Msln|MSLN_HUMAN|cleaved form|CAK1 antigen|Mesothelin|Mesothelin isoform 1 precursor
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:40 kDa (Mature form)/69 kDa (Precursor)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 273-407) of human Mesothelin (MSLN) protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-514
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rMSLN/8764] antibody to Mesothelin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Mesothelin
Clonality: Recombinant
Clone: rMSLN/8764
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human