Anti-MCM3 Mouse Monoclonal Antibody [clone: MCM3/6706]
Catalog # 77177-490
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Cervical cancer proto oncogene 5
- Antigen symbol:MCM3
- Clonality:Monoclonal
- Clone:MCM3/6706
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P25205
- Antigen synonyms:HCC5|DNA polymerase alpha holoenzyme-associated protein P1|HCC 5|hRlf beta subunit|Human cervical cancer proto oncogene 5|DNA polymerase alpha holoenzyme associated P1|DNA replication licensing factor mcm3|DNA polymerase alpha holoenzyme associated protein P1|DNA replication factor MCM3
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:115 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (aa 650-750) of human MCM3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-490
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MCM3/6706] antibody to MCM3 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: MCM3
Clonality: Monoclonal
Clone: MCM3/6706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human