Anti-liver FABP Mouse Monoclonal Antibody [clone: FABP1/4517]
Catalog # 77177-426
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:FABP 1
- Antigen symbol:liver FABP
- Clonality:Monoclonal
- Clone:FABP1/4517
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P07148
- Antigen synonyms:Fatty acid binding protein 1 liver|FABPL_HUMAN|FABPL|Fatty Acid Binding Protein 1|Fatty acid-binding protein|Fatty acid-binding protein liver|Fatty acid-binding protein 1|Fatty Acid Binding Protein|FABP1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:14 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human recombinant FABP1 protein fragment (around Amino Acids 1-127). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-426
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [FABP1/4517] antibody to liver FABP for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 2 to 4 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: liver FABP
Clonality: Monoclonal
Clone: FABP1/4517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human