Anti-liver FABP Rabbit Recombinant Antibody [clone: FABP1/8521R]
Catalog # 77177-424
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:FABP 1
- Antigen symbol:liver FABP
- Clonality:Recombinant
- Clone:FABP1/8521R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P07148
- Antigen synonyms:Fatty acid binding protein 1 liver|FABPL_HUMAN|FABPL|Fatty Acid Binding Protein 1|Fatty acid-binding protein|Fatty acid-binding protein liver|Fatty acid-binding protein 1|Fatty Acid Binding Protein|FABP1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:14 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human recombinant FABP1 protein fragment (around Amino Acids 1-127). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-424
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [FABP1/8521R] antibody to liver FABP for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: liver FABP
Clonality: Recombinant
Clone: FABP1/8521R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human