Anti-Leptin Receptor Mouse Monoclonal Antibody [clone: LEPR/4545]
Catalogue # 77177-406
Fournisseur: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Form:Liquid
- Gene ID:UniprotID# P48357
- Antigen synonyms:LEP-R|Db|CD295|HuB219|LEPR|Fa|LEPRD|CD295 antigen|LEP R
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:100 kDa/125 kDa (short/long form)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 335-425) of human LEPR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-406
- Packaging:Plastic vial
Spécifications
À propos de cet article
Mouse monoclonal [LEPR/4545] antibody to Leptin Receptor for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Leptin Receptor
Clonality: Monoclonal
Clone: LEPR/4545
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human