Anti-Ku80 Rabbit Recombinant Antibody [clone: XRCC5/8703R]
Catalog # 77177-364
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# P13010
- Antigen synonyms:ATP dependent DNA helicase 2 subunit 2|CTC85|CTC box binding factor 85 kDa|ATP-dependent DNA helicase 2 subunit 2|ATP dependent DNA helicase II 86 Kd subunit|ATP dependent DNA helicase II|ATP-dependent DNA helicase II 80 kDa subunit|CTC box-binding factor 85 kDa subunit|ATP dependent DNA helicase II 80 kDa subunit
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:80 - 86 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 300-500) of human XRCC5. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-364
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [XRCC5/8703R] antibody to Ku80 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Ku80
Clonality: Recombinant
Clone: XRCC5/8703R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human