Anti-Kallikrein 5 Mouse Monoclonal Antibody [clone: KLK5/3855]
Catalog # 77177-312
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# Q9Y337
- Antigen synonyms:Kallikrein-5|Kallikrein like 2|Kallikrein like protein 2|Kallikrein5|Kallikrein related peptidase 5|Kallikrein 5|Kallikrein L2|Kallikrein-like protein 2|KLK 5
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:33 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human KLK5 protein (around Amino Acids 1-200). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-312
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [KLK5/3855] antibody to Kallikrein 5 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Kallikrein 5
Clonality: Monoclonal
Clone: KLK5/3855
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human