Anti-IRF3 Mouse Monoclonal Antibody [clone: PCRP-IRF3-2F9]
77177-302
- Antibody type:Primary
- Antigen name:IIAE7
- Antigen symbol:IRF3
- Clonality:Monoclonal
- Clone:PCRP-IRF3-2F9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q14653
- Antigen synonyms:IRF3_HUMAN|Interferon regulatory factor 3|IRF3|MGC94729|IRF-3|IRF 3
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human IRF3 protein
- Purification:Protein A/G chromatography
- Cat. no.:77177-302
- Packaging:Plastic vial
Mouse monoclonal [PCRP-IRF3-2F9] antibody to IRF3 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: IRF3
Clonality: Monoclonal
Clone: PCRP-IRF3-2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human