Anti-INSM1 Rabbit Recombinant Antibody [clone: INSM1/8149R]
Catalog # 77177-290
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:IA1
- Antigen symbol:INSM1
- Clonality:Recombinant
- Clone:INSM1/8149R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q01101
- Antigen synonyms:INSM1|Zinc finger protein IA1|Insulinoma associated protein 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-300) corresponding to the N-terminus of human INSM1
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-290
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [INSM1/8149R] antibody to INSM1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: INSM1
Clonality: Recombinant
Clone: INSM1/8149R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human