Anti-IMP3 Rabbit Recombinant Antibody [clone: IMP3/8934R]
77177-280
- Antibody type:Primary
- Antigen name:BRMS2
- Antigen symbol:IMP3
- Clonality:Recombinant
- Clone:IMP3/8934R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NV31
- Antigen synonyms:DKFZp686F1078|CT98|DKFZp586L0118|hKOC|FLJ10968|IF2B3_HUMAN|Chromosome 15 open reading frame 12|Cancer/testis antigen 98|C15orf12
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human IMP3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-280
- Packaging:Plastic vial
Recombinant rabbit monoclonal [IMP3/8934R] antibody to IMP3 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: IMP3
Clonality: Recombinant
Clone: IMP3/8934R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human