Anti-hCG receptor/LHR Mouse Monoclonal Antibody [clone: LHCGR/7399]
Catalog # 77177-052
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen symbol:hCG receptor/LHR
- Clonality:Monoclonal
- Clone:LHCGR/7399
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P22888
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:78-85 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 100-300) of human LHCGR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77177-052
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [LHCGR/7399] antibody to hCG receptor/LHR for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: hCG receptor/LHR
Clonality: Monoclonal
Clone: LHCGR/7399
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human