Anti-Galectin 3 Mouse Monoclonal Antibody [clone: LGALS3/6583]
77176-890
- Form:Liquid
- Gene ID:UniprotID# P17931
- Antigen synonyms:Carbohydrate-binding protein 35|Galactose-specific lectin 3|GAL3|CBP35|GALBP|CBP 35|Carbohydrate binding protein 35|Gal-3|Galactoside-binding protein
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide from the C-terminus of human LGALS3 protein (aa 150-200). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77176-890
- Packaging:Plastic vial
Mouse monoclonal [LGALS3/6583] antibody to Galectin 3 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Galectin 3
Clonality: Monoclonal
Clone: LGALS3/6583
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human