Anti-BRG1 Rabbit Recombinant Antibody [clone: BRG1/8805R]
Catalog # 77175-970
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P51532
- Antigen synonyms:BAF 190|Brahma protein like 1|ATP-dependent helicase SMARCA4|BRG1 protein|BAF190|BRG1|BAF190A|BRG1-associated factor 190A|BRG1 associated factor 190A
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:185 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-400) of human SMARCA4 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:77175-970
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [BRG1/8805R] antibody to BRG1 for ELISA, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: BRG1
Clonality: Recombinant
Clone: BRG1/8805R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human