Anti-CTP synthase/CTPS Rabbit Monoclonal Antibody [clone: ARC2883]
Rabbit monoclonal [ARC2883] antibody to CTP synthase/CTPS for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: CTP synthase/CTPS
Clonality: Monoclonal
Clone: ARC2883
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together