Anti-SMAD5 (phospho Ser463 + Ser465) Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SMAD5 (phospho Ser463 + Ser465) for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: SMAD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77231-404
- Antigen symbol:
- SMAD5
- Antigen name:
- DKFZp781C1895
- Antigen synonyms:
- JV5 1|hSmad5|MADH5|Dwfc|JV5-1|MAD homolog 5|DKFZp781O1323|MAD, mothers against decapentaplegic homolog 5|MADH 5
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q99717
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Modification:
- phospho Ser463 + Ser465
- Immunogen:
- A synthetic phosphorylated peptide around S463 and S465 of human Phospho-Smad5-S463/S465 (NP_005894.3)
- Sequence:
- SSVS
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
Frequently Bought Together