Anti-AKT1 +AKT2 Rabbit Monoclonal Antibody [clone: ARC2651]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2651] antibody to AKT1 +AKT2 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: AKT1 +AKT2
Clonality: Monoclonal
Clone: ARC2651
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together