Anti-Myosin Phosphatase + PPP1R12B + PPP1R12C (phospho Thr696) Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Myosin Phosphatase + PPP1R12B + PPP1R12C (phospho Thr696) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Myosin Phosphatase + PPP1R12B + PPP1R12C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Related Information
Frequently Bought Together