Anti-SMARCA6 Rabbit Monoclonal Antibody [clone: ARC56706]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC56706] antibody to SMARCA6 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:2,000 to 1:20,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: SMARCA6
Clonality: Monoclonal
Clone: ARC56706
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together