Anti-p38 MAPK (phospho Thr180 + Tyr182) Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to p38 MAPK (phospho Thr180 + Tyr182) for WB with samples derived from Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: p38 MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Frequently Bought Together