Anti-PPP1R3C Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to PPP1R3C for IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ICC/IF
- Recommended Dilutions: IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: PPP1R3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together