Anti-Prostate Secretory Protein/PSP Rabbit Monoclonal Antibody [clone: ARC0912]
Rabbit monoclonal [ARC0912] antibody to Prostate Secretory Protein/PSP for WB and ICC/IF with samples derived from Human and Mouse.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Prostate Secretory Protein/PSP
Clonality: Monoclonal
Clone: ARC0912
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Frequently Bought Together