Anti-POU4F3 Goat Polyclonal Antibody
Catalog # 77067-342
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Brain specific homeobox/POU domain protein 3C
- Antigen symbol:POU4F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Brain-specific homeobox/POU domain protein 3C|DFNA15|Brain-3C|Brn-3C|DFNA 15|MGC138412|BRN 3C|class 4|BRN3C
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-EAAYREKNSKPE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human POU4F3 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77067-342
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to POU4F3 for ELISA and WB with samples derived from Human and Mouse.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: POU4F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse