Anti-VPS41 Goat Polyclonal Antibody
Catalog # 77064-230
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:HVPS41
- Antigen symbol:VPS41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Vacuolar protein sorting associated protein 41 homolog|Vacuolar protein sorting 41 homolog (S. cerevisiae)|Vacuolar protein sorting 41 (yeast homolog)|Vacuolar protein sorting 41 (yeast)|Vacuolar assembly protein 41|p|S53|vacuolar protein sorting-associated protein 41 homolog|HVSP41
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-DHLKKDSQNKTLLK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human VPS41 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77064-230
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to VPS41 for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IHC
Type: Primary
Antigen: VPS41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human