Anti-EYA1 Goat Polyclonal Antibody
Catalog # 77075-948
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:BOP
- Antigen symbol:EYA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:BOR|EYA1|EYA transcriptional coactivator and phosphatase 1|Eyes absent homolog 1|Eyes absent 1|EYA1_HUMAN|BOS1|Eyes absent homolog 1 (Drosophila)|Eyes absent 1 homolog
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-TDPTAEYSTIHSP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human EYA1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77075-948
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to EYA1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: EYA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human