Anti-CP110 Goat Polyclonal Antibody
77065-184
- Antibody type:Primary
- Antigen name:110 kDa centrosomal protein
- Antigen symbol:CP110
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Cep110|Centrosomal protein 110 kDa|DKFZp781G1416|FAN|Centrosomal protein of 110 kDa|Centrosomal protein CP110|CP110_HUMAN|CCP110|Centriolar coiled coil protein 110kDa
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CEKSLARIQEAS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CP110 (N terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77065-184
- Packaging:Plastic vial
Goat polyclonal antibody to CP110 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: CP110
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human