Anti-ADAM12 Goat Polyclonal Antibody
Catalog # 77069-702
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:A disintegrin and metalloproteinase domain 12
- Antigen symbol:ADAM12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Meltrin alpha|ADAM metallopeptidase domain 12|CAR10|ADAM12|ADA12_HUMAN|Disintegrin and metalloproteinase domain-containing protein 12|ADAM 12|MCMP|MCMPMltna
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-REFQRQGKDLEKVKQ
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ADAM12 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77069-702
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to ADAM12 for ELISA and WB with samples derived from Human, Mouse, Rat and Porcine.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: ADAM12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine