Anti-RAPSN Goat Polyclonal Antibody
Catalog # 77062-174
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:43 kD receptor associated protein of the synapse
- Antigen symbol:RAPSN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CMS1D|43 kDa postsynaptic protein|CMS1E|MGC3597|RAPSN|RAPSN_HUMAN|Acetylcholine receptor associated 43 kda protein|43 kDa receptor-associated protein of the synapse|Acetylcholine receptor-associated 43 kDa protein
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:YQSNQTEKALQVWTK-C
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human RAPSN (N terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77062-174
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to RAPSN for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: RAPSN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human