Anti-BLAP75 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to BLAP75 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:200 to 1:2000
Type: Primary
Antigen: BLAP75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 76852-976
- Antigen symbol:
- BLAP75
- Antigen name:
- BLAP75
- Antigen synonyms:
- FLJ12888|BLM associated protein 75 kDa|C9orf76|Homolog of yeast RecQ mediated genome instability 1|BLM associated polypeptide 75 kDa|BLM-associated protein of 75 kDa|FAAP75|Rmi1|RecQ-mediated genome instability protein 1
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9H9A7
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 350 of human RMI1 (NP_079221.2)
- Sequence:
- MNVTSIALRAETWLLAAWHVKVPPMWLEACINWIQEENNNVNLSQAQMNKQVFEQWLLTDLRDLEHPLLPDGILEIPKGELNGFYALQINSLVDVSQPAYSQIQKLRGKNTTNDLVTAEAQVTPKPWEAKPSRMLMLQLTDGIVQIQGMEYQPIPILHSDLPPGTKILIYGNISFRLGVLLLKPENVKVLGGEVDALLEEYAQEKVLARLIGEPDLVVSVIPNNSNENIPRVTDVLDPALGPSDEELLASLDENDELTANNDTSSERCFTTGSSSNTIPTRQSSFEPEFVISPRPKEEPSNLSIHVMDGELDDFSLEEALLLEETVQKEQMETKELQPLTFNRNADRSIE
- Form:
- Liquid
- Molecular weight:
- 75 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together