Anti-Neurogenin3/NGN-3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Neurogenin3 / NGN-3 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200
Type: Primary
Antigen: Neurogenin3/NGN-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 76850-592
- Antigen symbol:
- Neurogenin3/NGN-3
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9Y4Z2
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 214 of human NEUROG3 (NP_066279.2)
- Sequence:
- MTPQPSGAPTVQVTRETERSFPRASEDEVTCPTSAPPSPTRTRGNCAEAEEGGCRGAPRKLRARRGGRSRPKSELALSKQRRSRRKKANDRERNRMHNLNSALDALRGVLPTFPDDAKLTKIETLRFAHNYIWALTQTLRIADHSLYALEPPAPHCGELGSPGGSPGDWGSLYSPVSQAGSLSPAASLEERPGLLGATFSACLSPGSLAFSDFL
- Form:
- Liquid
- Molecular weight:
- 25 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.05% Proclin 300
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
Frequently Bought Together